Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TKTL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032136.4 | NP_115512.3 | 1 | 883 | ||
GenBank mRNA | AK057325.1 | - | 1 | 740 | ||
AK314517.1 | - | 1 | 791 | |||
AL136779.1 | - | 1 | 734 | |||
BC028707.2 | - | 1 | 826 | 4825223 | ||
BC125101.1 | - | 1 | 756 | 40118304 | ||
BC142943.1 | - | 1 | 731 | 8860325 | ||
CR533560.1 | - | 1 | 722 |