Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACCS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127219.1 | NP_001120691.1 | 7 | 755 | ||
NM_032592.3 | NP_115981.1 | 7 | 1051 | |||
XM_005253170.3 | XP_005253227.2 | 7 | 1194 | |||
XM_005253172.2 | XP_005253229.1 | 4 | 282 | |||
XM_006718348.2 | XP_006718411.1 | 6, 7 | 503 | |||
XM_011520406.2 | XP_011518708.1 | 7 | 1197 | |||
XM_011520407.1 | XP_011518709.1 | 7 | 547 | |||
XM_011520410.2 | XP_011518712.1 | 7 | 1213 | |||
XM_017018421.1 | XP_016873910.1 | 6, 7 | 1154 | |||
GenBank mRNA | AK057649.1 | - | 7 | 1045 | ||
AK302515.1 | - | 6, 7 | 518 | |||
AK304074.1 | - | 6 | 917 | |||
AL833533.1 | - | 1, 2 | 73 | |||
AY026508.1 | - | 7 | 1006 | |||
BC020197.1 | - | 7 | 1025 | 3632065 | ||
KU178811.1 | - | 6 | 611 | |||
KU178812.1 | - | 6 | 611 | |||
KU178813.1 | - | 5 | 551 |