Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRMT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304458.1 | NP_001291387.1 | 2 | 481 | ||
NM_138364.3 | NP_612373.2 | 3 | 630 | |||
XM_017008823.1 | XP_016864312.1 | 3 | 406 | |||
XM_017008824.1 | XP_016864313.1 | 3 | 406 | |||
XM_017008825.1 | XP_016864314.1 | 2 | 392 | |||
XM_017008826.1 | XP_016864315.1 | 2 | 355 | |||
GenBank mRNA | AK096703.1 | - | 2 | 376 | ||
AK292904.1 | - | 3 | 563 | |||
BC021250.2 | - | 3 | 385 | |||
BC064403.1 | - | 3 | 529 | 5547611 |