Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRMT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257994.1 | NP_001244923.1 | 12 | 2532 | ||
NM_022727.5 | NP_073564.3 | 12 | 2510 | |||
NM_182984.4 | NP_892029.2 | 13 | 2386 | |||
XM_011530139.2 | XP_011528441.1 | 13 | 2371 | |||
XM_011530141.1 | XP_011528443.1 | 11 | 1746 | |||
XM_011530142.2 | XP_011528444.1 | 9 | 1404 | |||
XM_017028770.1 | XP_016884259.1 | 12 | 2574 | |||
GenBank mRNA | AK025106.1 | - | 11 | 1906 | ||
AK057029.1 | - | 12 | 2482 | |||
BC013352.2 | - | 12 | 2155 | 4054100 | ||
BC017184.2 | - | 13 | 2272 | 3347641 | ||
BC108251.1 | - | 12 | 2190 | 3852106 |