Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPHK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142601.1 | NP_001136073.1 | 3 | 532 | ||
NM_001142602.1 | NP_001136074.1 | 2 | 577 | |||
NM_021972.3 | NP_068807.2 | 3 | 574 | |||
NM_182965.2 | NP_892010.2 | 3 | 895 | |||
XM_005257766.2 | XP_005257823.1 | 3 | 887 | |||
GenBank mRNA | AF200328.1 | - | 2 | 158 | ||
AF238083.1 | - | 3 | 476 | |||
AF266756.1 | - | 2 | 409 | |||
AJ245504.1 | - | 1 | 131 | |||
AK022402.1 | - | 3 | 532 | |||
AK023393.1 | - | 2 | 489 | |||
AK095578.1 | - | 2 | 1217 | |||
AK122781.1 | - | 5 | 949 | |||
AK225250.1 | - | 3 | 495 | |||
AK225258.1 | - | 3 | 495 | |||
AK292294.1 | - | 3 | 619 | |||
AK292614.1 | - | 2 | 493 | |||
BC008040.2 | - | 3 | 469 | 3831657 | ||
BC009419.1 | - | 3 | 562 | 3506924 | ||
BC014439.1 | - | 3 | 832 | 4871343 | ||
BC030553.1 | - | 3 | 854 | 5213270 | ||
DC384776.1 | - | 2 | 505 |