Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278282.1 | NP_001265211.1 | 7 | 1396 | ||
NM_001278283.1 | NP_001265212.1 | 8 | 1476 | |||
NM_001278285.1 | NP_001265214.1 | 5 | 1304 | |||
NM_001278286.1 | NP_001265215.1 | 6 | 1320 | |||
NM_001278288.1 | NP_001265217.1 | 5 | 1311 | |||
NM_001278289.1 | NP_001265218.1 | 7 | 1460 | |||
NM_001318926.1 | NP_001305855.1 | 7 | 1445 | |||
NM_001318927.1 | NP_001305856.1 | 7 | 1489 | |||
NM_001318928.1 | NP_001305857.1 | 7 | 1340 | |||
NM_001318929.1 | NP_001305858.1 | 6 | 1387 | |||
NM_001318930.1 | NP_001305859.1 | 6 | 1380 | |||
NM_001318931.1 | NP_001305860.1 | 6 | 1342 | |||
NM_006916.2 | NP_008847.1 | 6 | 1365 | |||
NM_199229.2 | NP_954699.1 | 6 | 1391 | |||
XM_006712677.3 | XP_006712740.1 | 7 | 1338 | |||
GenBank mRNA | AB075523.1 | - | 1 | 16 | ||
AJ224326.1 | - | 5 | 1133 | |||
AK056028.1 | - | 6 | 1302 | |||
AK093658.1 | - | 8 | 1407 | |||
AK096199.1 | - | 7 | 1227 | |||
AK303184.1 | - | 5 | 1231 | |||
AK308741.1 | - | 6 | 1229 | |||
BC005148.2 | - | 6 | 1299 | 3505010 | ||
BC016764.1 | - | 6 | 1328 | 4107315 | ||
BC072401.1 | - | 6 | 1300 | 6047438 |
Gene Symbol: RPEL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143909.1 | NP_001137381.1 | 1 | 1409 | ||
GenBank mRNA | BC157875.1 | - | 1 | 1410 | 9057093 | |
BC171885.1 | - | 1 | 1411 | 9054539 |