Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000927.4 | NP_000918.2 | 7 | 1011 | ||
GenBank mRNA | AB208970.1 | - | 5 | 483 | ||
AF016535.1 | - | 6 | 637 | |||
AK290159.1 | - | 7 | 1012 | |||
BC130424.1 | - | 6 | 843 | 40146455 | ||
EU852583.1 | - | 5 | 518 | |||
M14758.1 | - | 7 | 942 |