Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000927.4 | NP_000918.2 | 10 | 1384 | ||
GenBank mRNA | AB208970.1 | - | 8 | 854 | ||
AF016535.1 | - | 9 | 1010 | |||
AK290159.1 | - | 10 | 1385 | |||
BC130424.1 | - | 9 | 1216 | 40146455 | ||
BQ882401.1 | - | 2 | 157 | |||
EU852583.1 | - | 8 | 891 | |||
EU854148.1 | - | 7 | 699 | |||
M14758.1 | - | 10 | 1315 |