Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025091.1 | NP_001020262.1 | 10 | 941 | ||
NM_001090.2 | NP_001081.1 | 9 | 827 | |||
GenBank mRNA | AB209564.1 | - | 2 | 104 | ||
AF027302.1 | - | 9 | 826 | |||
AK298061.1 | - | 10 | 884 | |||
AK307551.1 | - | 10 | 895 | |||
AL832430.1 | - | 4 | 371 | |||
BC034488.2 | - | 10 | 886 | 4826422 | ||
BC112923.1 | - | 10 | 874 | 7470309 |