Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002858.3 | NP_002849.1 | 23 | 2344 | ||
XM_006710802.2 | XP_006710865.2 | 24 | 2707 | |||
XM_011541877.1 | XP_011540179.1 | 16 | 1590 | |||
XM_017001952.1 | XP_016857441.1 | 23 | 2885 | |||
XM_017001953.1 | XP_016857442.1 | 23 | 2265 | |||
XM_017001954.1 | XP_016857443.1 | 22 | 2325 | |||
GenBank mRNA | AK126440.1 | - | 2 | 2069 | ||
AK302714.1 | - | 22 | 2269 | |||
BC068509.1 | - | 19 | 1975 | 5265201 | ||
BX648715.1 | - | 14 | 3092 | |||
M81182.1 | - | 23 | 2243 | |||
X58528.1 | - | 23 | 2269 |