Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005502.3 | NP_005493.2 | 7 | 1090 | ||
XM_005251773.2 | XP_005251830.1 | 7 | 999 | |||
XM_005251776.2 | XP_005251833.1 | 6 | 839 | |||
XM_011518339.2 | XP_011516641.1 | 8 | 1074 | |||
XM_011518340.2 | XP_011516642.1 | 9 | 1289 | |||
XM_011518341.2 | XP_011516643.1 | 8 | 1074 | |||
XM_011518342.2 | XP_011516644.1 | 5 | 777 | |||
XM_011518344.2 | XP_011516646.1 | 8 | 1074 | |||
XM_017014378.1 | XP_016869867.1 | 8 | 1272 | |||
XM_017014379.1 | XP_016869868.1 | 8 | 1680 | |||
XM_017014380.1 | XP_016869869.1 | 8 | 1006 | |||
XM_017014381.1 | XP_016869870.1 | 8 | 1072 | |||
XM_017014382.1 | XP_016869871.1 | 6 | 1537 | |||
GenBank mRNA | AB055982.1 | - | 6 | 686 | ||
AB445477.1 | - | 6 | 686 | |||
AF165281.1 | - | 5 | 626 | |||
AF285167.1 | - | 7 | 976 | |||
AJ012376.1 | - | 5 | 626 | |||
AK024328.1 | - | 7 | 999 | |||
BC146856.1 | - | 7 | 900 | 9056784 |