Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005689.2 | NP_005680.1 | 3 | 1151 | ||
GenBank mRNA | AF076775.1 | - | 3 | 834 | ||
AF308472.1 | - | 3 | 1140 | |||
AJ289233.2 | - | 3 | 1111 | |||
AK057026.1 | - | 3 | 870 | |||
AK172812.1 | - | 3 | 501 | |||
BC000559.2 | - | 3 | 1102 | 3162881 |