Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204813.1 | NP_001191742.1 | 5 | 1575 | ||
NM_002526.3 | NP_002517.1 | 5 | 1575 | |||
GenBank mRNA | AK075008.1 | - | 5 | 1050 | ||
AK075083.1 | - | 2 | 277 | |||
AK314661.1 | - | 5 | 1089 | |||
BC065937.1 | - | 5 | 1044 | 6163101 | ||
DQ186653.1 | - | 5 | 1038 | |||
X55740.1 | - | 5 | 1068 |