Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320653.1 | NP_001307582.1 | 15, 16 | 2188 | ||
NM_002532.5 | NP_002523.2 | 15, 16 | 2140 | |||
XM_011523893.2 | XP_011522195.1 | 16, 17 | 2238 | |||
XM_017024694.1 | XP_016880183.1 | 16, 17 | 2190 | |||
GenBank mRNA | AK298158.1 | - | 16, 17 | 1965 | ||
AK308142.1 | - | 15, 16 | 1953 | |||
BC000335.2 | - | 15, 16 | 2053 | 2822595 | ||
CX870338.1 | - | 2, 3 | 223 | |||
Y08612.2 | - | 15, 16 | 2099 |