Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NVL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243146.1 | NP_001230075.1 | 4 | 354 | ||
NM_001243147.1 | NP_001230076.1 | 6 | 486 | |||
NM_002533.3 | NP_002524.2 | 7 | 759 | |||
NM_206840.2 | NP_996671.1 | 6 | 685 | |||
XM_011544196.1 | XP_011542498.1 | 7 | 742 | |||
XM_011544198.1 | XP_011542500.1 | 6 | 697 | |||
XM_011544199.1 | XP_011542501.1 | 6 | 651 | |||
XM_011544200.1 | XP_011542502.1 | 5 | 625 | |||
XM_017001378.1 | XP_016856867.1 | 8 | 851 | |||
XM_017001379.1 | XP_016856868.1 | 8 | 851 | |||
XM_017001380.1 | XP_016856869.1 | 8 | 851 | |||
XM_017001381.1 | XP_016856870.1 | 8 | 848 | |||
XM_017001382.1 | XP_016856871.1 | 7 | 781 | |||
XM_017001383.1 | XP_016856872.1 | 7 | 781 | |||
XM_017001384.1 | XP_016856873.1 | 8 | 851 | |||
XM_017001385.1 | XP_016856874.1 | 8 | 851 | |||
XM_017001386.1 | XP_016856875.1 | 5 | 625 | |||
GenBank mRNA | AK297070.1 | - | 7 | 718 | ||
AK297396.1 | - | 4 | 305 | |||
AK298244.1 | - | 6 | 435 | |||
AK298826.1 | - | 6 | 637 | |||
BC012105.1 | - | 5 | 346 | 4636004 | ||
BQ883523.1 | - | 2 | 280 | |||
U68140.1 | - | 7 | 753 | |||
U78772.1 | - | 6 | 666 |