Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDGFD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025208.4 | NP_079484.1 | 6 | 1341 | ||
NM_033135.3 | NP_149126.1 | 6 | 1323 | |||
GenBank mRNA | AB033832.1 | - | 6 | 1125 | ||
AF113216.1 | - | 6 | 1075 | |||
AF335584.1 | - | 6 | 1150 | |||
AF336376.1 | - | 6 | 1144 | |||
AK292801.1 | - | 6 | 1149 | |||
AY027517.1 | - | 6 | 1061 | |||
AY027518.1 | - | 6 | 1043 | |||
AY359116.1 | - | 6 | 982 | |||
BC030645.2 | - | 6 | 1403 | 4824526 |