Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001823.4 | NP_001814.2 | 6, 7 | 887 | ||
XM_017020951.1 | XP_016876440.1 | 5, 6 | 933 | |||
GenBank mRNA | AK290101.1 | - | 6, 7 | 858 | ||
AK298198.1 | - | 6, 7 | 756 | |||
AK312282.1 | - | 6, 7 | 859 | |||
BC001190.1 | - | 6, 7 | 846 | 3354679 | ||
BC004914.2 | - | 6, 7 | 838 | 3537262 | ||
BC008323.1 | - | 6, 7 | 844 | 3358085 | ||
BC010002.2 | - | 6, 7 | 853 | 4135106 | ||
BC019259.1 | - | 6, 7 | 842 | 3503057 | ||
BC019281.1 | - | 6, 7 | 852 | 2823530 | ||
CR542268.1 | - | 5, 6 | 781 | |||
EU794629.1 | - | 6, 7 | 842 | |||
FJ224337.1 | - | 5, 6 | 781 | |||
KU177977.1 | - | 5, 6 | 781 | |||
KU177978.1 | - | 4, 5 | 853 | |||
L47647.1 | - | 6, 7 | 824 | |||
M16364.1 | - | 5, 6 | 789 | |||
M16451.1 | - | 5, 6 | 804 |