Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001823.4 | NP_001814.2 | 7 | 1026 | ||
XM_017020951.1 | XP_016876440.1 | 6 | 1072 | |||
GenBank mRNA | AK290101.1 | - | 7 | 997 | ||
AK298198.1 | - | 7 | 895 | |||
AK312282.1 | - | 7 | 998 | |||
BC001190.1 | - | 7 | 985 | 3354679 | ||
BC004914.2 | - | 7 | 977 | 3537262 | ||
BC008323.1 | - | 7 | 983 | 3358085 | ||
BC010002.2 | - | 7 | 992 | 4135106 | ||
BC019259.1 | - | 7 | 981 | 3503057 | ||
BC019281.1 | - | 7 | 991 | 2823530 | ||
CR542268.1 | - | 6 | 920 | |||
EU794629.1 | - | 7 | 981 | |||
FJ224337.1 | - | 6 | 920 | |||
KU177977.1 | - | 6 | 920 | |||
KU177978.1 | - | 5 | 992 | |||
L47647.1 | - | 7 | 963 | |||
M16364.1 | - | 6 | 928 | |||
M16451.1 | - | 6 | 943 |