Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NME3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002513.2 | NP_002504.2 | 4, 5 | 567 | ||
XM_011522504.2 | XP_011520806.1 | 4, 5 | 614 | |||
GenBank mRNA | AK308663.1 | - | 4, 5 | 388 | ||
BC000250.1 | - | 4, 5 | 567 | 3357930 | ||
BG911570.1 | - | 4, 5 | 391 |