Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204402.1 | NP_001191331.1 | 3 | 636 | ||
NM_002764.3 | NP_002755.1 | 6 | 1044 | |||
GenBank mRNA | AK223169.1 | - | 6 | 861 | ||
AK297968.1 | - | 7 | 882 | |||
AK307748.1 | - | 5 | 860 | |||
AK316467.1 | - | 3 | 550 | |||
BC001605.1 | - | 6 | 939 | 3542584 | ||
D00860.1 | - | 6 | 961 | |||
X15331.1 | - | 6 | 905 |