Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSNK1G2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319.6 | NP_001310.3 | 2, 3 | 708 | ||
XM_005259498.1 | XP_005259555.1 | 2, 3 | 720 | |||
XM_005259499.3 | XP_005259556.1 | 2, 3 | 527 | |||
XM_005259500.1 | XP_005259557.1 | 2, 3 | 720 | |||
XM_017026297.1 | XP_016881786.1 | 2, 3 | 594 | |||
GenBank mRNA | AB451278.1 | - | 1, 2 | 185 | ||
AB451410.1 | - | 1, 2 | 185 | |||
BC018693.2 | - | 2, 3 | 681 | 3010717 | ||
BC018699.2 | - | 2, 3 | 681 | 3010717 | ||
BC020972.1 | - | 1, 2 | 238 | 3851613 | ||
BT009922.1 | - | 1, 2 | 185 | |||
U89896.1 | - | 1, 2 | 424 |