Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287135.1 | NP_001274064.1 | 3 | 517 | ||
NM_001287136.1 | NP_001274065.1 | 2 | 270 | |||
NM_001287137.1 | NP_001274066.1 | 2 | 270 | |||
NM_012395.3 | NP_036527.1 | 2 | 341 | |||
XM_005250436.3 | XP_005250493.1 | 4 | 316 | |||
XM_005250438.3 | XP_005250495.1 | 2 | 263 | |||
XM_005250439.2 | XP_005250496.1 | 3 | 1074 | |||
XM_011516306.2 | XP_011514608.1 | 3 | 885 | |||
XM_011516307.2 | XP_011514609.1 | 3 | 885 | |||
XM_017012320.1 | XP_016867809.1 | 4 | 449 | |||
XM_017012321.1 | XP_016867810.1 | 2 | 178 | |||
XM_017012322.1 | XP_016867811.1 | 2 | 820 | |||
GenBank mRNA | AB020641.1 | - | 2 | 341 | ||
AF119833.1 | - | 3 | 389 | |||
AK289782.1 | - | 2 | 199 | |||
AK295086.1 | - | 2 | 253 | |||
AK296406.1 | - | 2 | 224 | |||
AK297974.1 | - | 3 | 473 | |||
AK307484.1 | - | 2 | 224 | |||
AK316026.1 | - | 2 | 224 | |||
BC136476.1 | - | 2 | 411 | 9020465 | ||
BC136477.1 | - | 2 | 296 | 9020466 | ||
BC152388.1 | - | 2 | 200 | |||
BC152436.1 | - | 2 | 341 | 8862558 | ||
BC167152.1 | - | 2 | 341 | 8862547 | ||
BC167156.1 | - | 2 | 341 | 8862570 | ||
DC325068.1 | - | 2 | 273 | |||
DC420282.1 | - | 3 | 508 | |||
DQ655962.2 | - | 4 | 297 | |||
DQ655963.2 | - | 3 | 251 | |||
DQ655965.2 | - | 4 | 323 | |||
DQ655966.2 | - | 5 | 352 | |||
DQ656060.2 | - | 6 | 490 |