Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287135.1 | NP_001274064.1 | 14 | 1688 | ||
NM_001287136.1 | NP_001274065.1 | 13 | 1441 | |||
NM_001287137.1 | NP_001274066.1 | 12 | 1346 | |||
NM_012395.3 | NP_036527.1 | 13 | 1512 | |||
XM_005250436.3 | XP_005250493.1 | 15 | 1487 | |||
XM_005250438.3 | XP_005250495.1 | 13 | 1434 | |||
XM_005250439.2 | XP_005250496.1 | 14 | 2245 | |||
XM_011516306.2 | XP_011514608.1 | 14 | 2056 | |||
XM_017012320.1 | XP_016867809.1 | 15 | 1620 | |||
XM_017012321.1 | XP_016867810.1 | 13 | 1349 | |||
XM_017012322.1 | XP_016867811.1 | 13 | 1991 | |||
GenBank mRNA | AB020641.1 | - | 13 | 1512 | ||
AF119833.1 | - | 14 | 1560 | |||
AK289782.1 | - | 13 | 1370 | |||
AK295086.1 | - | 13 | 1424 | |||
AK296406.1 | - | 12 | 1300 | |||
AK297974.1 | - | 14 | 1644 | |||
AK307484.1 | - | 13 | 1395 | |||
AK316026.1 | - | 13 | 1395 | |||
BC136476.1 | - | 13 | 1582 | 9020465 | ||
BC136477.1 | - | 13 | 1467 | 9020466 | ||
BC152436.1 | - | 13 | 1512 | 8862558 | ||
BC167152.1 | - | 13 | 1512 | 8862547 | ||
BC167156.1 | - | 13 | 1512 | 8862570 |