Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FGFR1 SIRNA Assays ›
Gene Symbol:
FGFR1
Gene Name:
fibroblast growth factor receptor 1
Gene Aliases:
BFGFR, CD331, CEK, ECCL, FGFBR, FGFR-1, FLG, FLT-2, FLT2, HBGFR, HH2, HRTFDS, KAL2, N-SAM, OGD, bFGF-R-1
Chromosome Location:
Chr.8: 38411138 - 38468834 on Build GRCh38
UniGene ID:
Hs.264887
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
118294
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FGFR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001174063.1
NP_001167534.1
5
1545
NM_001174064.1
NP_001167535.1
6
1613
NM_001174065.1
NP_001167536.1
5
859
NM_001174066.1
NP_001167537.1
4
598
NM_001174067.1
NP_001167538.1
6
1019
NM_015850.3
NP_056934.2
5
1539
NM_023105.2
NP_075593.1
4
1278
NM_023106.2
NP_075594.1
4
1272
NM_023110.2
NP_075598.2
5
1545
XM_006716303.2
XP_006716366.1
5
1346
XM_006716304.1
XP_006716367.1
5
862
XM_006716306.2
XP_006716369.1
5
1340
XM_006716307.1
XP_006716370.1
5
856
XM_006716309.3
XP_006716372.1
5
1316
XM_006716310.2
XP_006716373.1
4
1079
XM_006716311.1
XP_006716374.1
4
477
XM_006716312.1
XP_006716375.1
4
547
XM_006716313.2
XP_006716376.1
4
1073
XM_006716314.1
XP_006716377.1
4
551
XM_011544443.2
XP_011542745.1
5
851
XM_011544444.1
XP_011542746.1
5
840
XM_011544445.2
XP_011542747.1
5
859
XM_011544446.2
XP_011542748.1
5
846
XM_011544447.2
XP_011542749.1
5
846
XM_011544448.1
XP_011542750.1
4
450
XM_011544449.1
XP_011542751.1
4
444
XM_011544450.2
XP_011542752.1
4
444
XM_011544451.1
XP_011542753.1
3
313
XM_011544452.2
XP_011542754.1
5
934
XM_017013219.1
XP_016868708.1
5
847
XM_017013220.1
XP_016868709.1
5
840
XM_017013221.1
XP_016868710.1
6
1055
XM_017013222.1
XP_016868711.1
5
1346
XM_017013223.1
XP_016868712.1
5
856
XM_017013224.1
XP_016868713.1
5
1340
XM_017013225.1
XP_016868714.1
5
823
XM_017013226.1
XP_016868715.1
4
450
XM_017013227.1
XP_016868716.1
4
444
XM_017013228.1
XP_016868717.1
4
1073
XM_017013230.1
XP_016868719.1
3
404
XM_017013231.1
XP_016868720.1
5
944
GenBank mRNA
AB208919.1
-
6
1400
AK222718.1
-
5
1338
AK223126.1
-
4
1073
AK291754.1
-
5
738
AK292470.1
-
4
598
AK309947.1
-
6
898
BC015035.1
-
6
1273
3911101
BC018128.1
-
5
1303
3896359
BC091494.1
-
3
409
30387634
BX649167.1
-
1
2887
EU826596.1
-
3
330
FJ809916.1
-
5
1548
FJ809917.1
-
5
1548
KU178066.1
-
4
603
KU178067.1
-
3
336
KU178068.1
-
3
330
KU178069.1
-
3
336
KU178070.1
-
3
330
M34185.1
-
4
591
M34186.1
-
4
856
M34187.1
-
4
752
M34188.1
-
4
852
M34641.1
-
4
606
M37722.1
-
4
558
M60485.1
-
6
1268
M63887.1
-
4
603
M63888.1
-
4
603
M63889.1
-
4
480
X51803.1
-
5
720
X52833.1
-
4
690
X57118.1
-
5
741
X57119.1
-
4
498
X57120.1
-
5
759
X57121.1
-
5
765
X57122.1
-
4
492
X66945.1
-
5
1329
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
MAPK cascade
skeletal system development
angiogenesis
ureteric bud development
in utero embryonic development
organ induction
neuron migration
positive regulation of mesenchymal cell proliferation
chondrocyte differentiation
transcription, DNA-templated
protein phosphorylation
sensory perception of sound
positive regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
positive regulation of phospholipase activity
positive regulation of phospholipase C activity
positive regulation of neuron projection development
regulation of phosphatidylinositol 3-kinase signaling
positive regulation of phosphatidylinositol 3-kinase signaling
cell migration
peptidyl-tyrosine phosphorylation
stem cell population maintenance
motogenic signaling involved in postnatal olfactory bulb interneuron migration
ventricular zone neuroblast division
embryonic limb morphogenesis
midbrain development
neuron projection development
fibroblast growth factor receptor signaling pathway involved in orbitofrontal cortex development
phosphatidylinositol-3-phosphate biosynthetic process
inner ear morphogenesis
outer ear morphogenesis
middle ear morphogenesis
chordate embryonic development
positive regulation of MAP kinase activity
positive regulation of MAPK cascade
positive regulation of GTPase activity
regulation of cell differentiation
positive regulation of neuron differentiation
negative regulation of osteoblast differentiation
positive regulation of cell cycle
protein autophosphorylation
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
paraxial mesoderm development
regulation of lateral mesodermal cell fate specification
cell maturation
skeletal system morphogenesis
mesenchymal cell differentiation
regulation of sensory perception of pain
positive regulation of cardiac muscle cell proliferation
auditory receptor cell development
branching involved in salivary gland morphogenesis
lung-associated mesenchyme development
regulation of branching involved in salivary gland morphogenesis by mesenchymal-epithelial signaling
regulation of stem cell proliferation
positive regulation of MAPKKK cascade by fibroblast growth factor receptor signaling pathway
positive regulation of endothelial cell chemotaxis to fibroblast growth factor
regulation of extrinsic apoptotic signaling pathway in absence of ligand
glycoprotein binding
protein tyrosine kinase activity
fibroblast growth factor-activated receptor activity
Ras guanyl-nucleotide exchange factor activity
protein binding
ATP binding
heparin binding
1-phosphatidylinositol-3-kinase activity
fibroblast growth factor binding
protein complex binding
identical protein binding
protein homodimerization activity
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
cell adhesion molecule binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6c48f4b768-zvmr9:80/10.239.162.158:80.
git-commit: a2f5e26549390d543f5d2d1fbad8092df65fbda4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.10.0-2024.06.23-1.0