Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FGFR1 SIRNA Assays ›
Gene Symbol:
FGFR1
Gene Name:
fibroblast growth factor receptor 1
Gene Aliases:
BFGFR, CD331, CEK, ECCL, FGFBR, FGFR-1, FLG, FLT-2, FLT2, HBGFR, HH2, HRTFDS, KAL2, N-SAM, OGD, bFGF-R-1
Chromosome Location:
Chr.8: 38411138 - 38468834 on Build GRCh38
UniGene ID:
Hs.264887
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
118295
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FGFR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001174063.1
NP_001167534.1
6
1591
NM_001174064.1
NP_001167535.1
7
1659
NM_001174065.1
NP_001167536.1
6
905
NM_001174066.1
NP_001167537.1
5
644
NM_001174067.1
NP_001167538.1
7
1065
NM_015850.3
NP_056934.2
6
1585
NM_023105.2
NP_075593.1
5
1324
NM_023106.2
NP_075594.1
5
1318
NM_023110.2
NP_075598.2
6
1591
XM_006716303.2
XP_006716366.1
6
1392
XM_006716304.1
XP_006716367.1
6
908
XM_006716306.2
XP_006716369.1
6
1386
XM_006716307.1
XP_006716370.1
6
902
XM_006716309.3
XP_006716372.1
6
1362
XM_006716310.2
XP_006716373.1
5
1125
XM_006716311.1
XP_006716374.1
5
523
XM_006716312.1
XP_006716375.1
5
593
XM_006716313.2
XP_006716376.1
5
1119
XM_006716314.1
XP_006716377.1
5
597
XM_011544443.2
XP_011542745.1
6
897
XM_011544444.1
XP_011542746.1
6
886
XM_011544445.2
XP_011542747.1
6
905
XM_011544446.2
XP_011542748.1
6
892
XM_011544447.2
XP_011542749.1
6
892
XM_011544448.1
XP_011542750.1
5
496
XM_011544449.1
XP_011542751.1
5
490
XM_011544450.2
XP_011542752.1
5
490
XM_011544451.1
XP_011542753.1
4
359
XM_011544452.2
XP_011542754.1
6
980
XM_017013219.1
XP_016868708.1
6
893
XM_017013220.1
XP_016868709.1
6
886
XM_017013221.1
XP_016868710.1
7
1101
XM_017013222.1
XP_016868711.1
6
1392
XM_017013223.1
XP_016868712.1
6
902
XM_017013224.1
XP_016868713.1
6
1386
XM_017013225.1
XP_016868714.1
6
869
XM_017013226.1
XP_016868715.1
5
496
XM_017013227.1
XP_016868716.1
5
490
XM_017013228.1
XP_016868717.1
5
1119
XM_017013230.1
XP_016868719.1
4
450
XM_017013231.1
XP_016868720.1
6
990
GenBank mRNA
AB208919.1
-
7
1446
AK222718.1
-
6
1384
AK223126.1
-
5
1119
AK291754.1
-
6
784
AK292470.1
-
5
644
AK309947.1
-
7
944
BC015035.1
-
7
1319
3911101
BC018128.1
-
6
1349
3896359
BC091494.1
-
4
455
30387634
BX649167.1
-
2
2933
EU826596.1
-
4
376
EU826597.1
-
4
382
FJ809916.1
-
6
1594
FJ809917.1
-
6
1594
KU178066.1
-
5
649
KU178067.1
-
4
382
KU178068.1
-
4
376
KU178069.1
-
4
382
KU178070.1
-
4
376
M34185.1
-
5
637
M34186.1
-
5
902
M34187.1
-
5
798
M34188.1
-
5
898
M34641.1
-
5
652
M37722.1
-
5
604
M60485.1
-
7
1314
M63887.1
-
5
649
M63888.1
-
5
649
M63889.1
-
5
526
X51803.1
-
6
766
X52833.1
-
5
736
X57118.1
-
6
787
X57119.1
-
5
544
X57120.1
-
6
805
X57121.1
-
6
811
X57122.1
-
5
538
X66945.1
-
6
1375
Y00665.1
-
2
49
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_024146, XM_006253323, XM_006253325, XM_006253327, XM_006253324, and others.
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
MAPK cascade
skeletal system development
angiogenesis
ureteric bud development
in utero embryonic development
organ induction
neuron migration
positive regulation of mesenchymal cell proliferation
chondrocyte differentiation
transcription, DNA-templated
protein phosphorylation
sensory perception of sound
positive regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
positive regulation of phospholipase activity
positive regulation of phospholipase C activity
positive regulation of neuron projection development
regulation of phosphatidylinositol 3-kinase signaling
positive regulation of phosphatidylinositol 3-kinase signaling
cell migration
peptidyl-tyrosine phosphorylation
stem cell population maintenance
motogenic signaling involved in postnatal olfactory bulb interneuron migration
ventricular zone neuroblast division
embryonic limb morphogenesis
midbrain development
neuron projection development
fibroblast growth factor receptor signaling pathway involved in orbitofrontal cortex development
phosphatidylinositol-3-phosphate biosynthetic process
inner ear morphogenesis
outer ear morphogenesis
middle ear morphogenesis
chordate embryonic development
positive regulation of MAP kinase activity
positive regulation of MAPK cascade
positive regulation of GTPase activity
regulation of cell differentiation
positive regulation of neuron differentiation
negative regulation of osteoblast differentiation
positive regulation of cell cycle
protein autophosphorylation
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
paraxial mesoderm development
regulation of lateral mesodermal cell fate specification
cell maturation
skeletal system morphogenesis
mesenchymal cell differentiation
regulation of sensory perception of pain
positive regulation of cardiac muscle cell proliferation
auditory receptor cell development
branching involved in salivary gland morphogenesis
lung-associated mesenchyme development
regulation of branching involved in salivary gland morphogenesis by mesenchymal-epithelial signaling
regulation of stem cell proliferation
positive regulation of MAPKKK cascade by fibroblast growth factor receptor signaling pathway
positive regulation of endothelial cell chemotaxis to fibroblast growth factor
regulation of extrinsic apoptotic signaling pathway in absence of ligand
glycoprotein binding
protein tyrosine kinase activity
fibroblast growth factor-activated receptor activity
Ras guanyl-nucleotide exchange factor activity
protein binding
ATP binding
heparin binding
1-phosphatidylinositol-3-kinase activity
fibroblast growth factor binding
protein complex binding
identical protein binding
protein homodimerization activity
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
cell adhesion molecule binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6c48f4b768-sx5kz:80/10.239.163.240:80.
git-commit: a2f5e26549390d543f5d2d1fbad8092df65fbda4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.10.0-2024.06.23-1.0