Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001369.2 | NP_001360.1 | 14 | 1865 | ||
XM_005248262.3 | XP_005248319.2 | 14 | 1948 | |||
XM_017009177.1 | XP_016864666.1 | 14 | 1948 | |||
XM_017009178.1 | XP_016864667.1 | 14 | 2751 | |||
XM_017009179.1 | XP_016864668.1 | 8 | 1045 | |||
XM_017009180.1 | XP_016864669.1 | 14 | 1948 | |||
XM_017009181.1 | XP_016864670.1 | 14 | 1948 | |||
XM_017009182.1 | XP_016864671.1 | 14 | 1948 | |||
XM_017009183.1 | XP_016864672.1 | 14 | 1948 | |||
XM_017009184.1 | XP_016864673.1 | 14 | 1948 | |||
XM_017009187.1 | XP_016864676.1 | 14 | 1948 | |||
GenBank mRNA | AY045575.1 | - | 14 | 1865 |