Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNC1LI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323955.1 | NP_001310884.1 | 3, 4 | 651 | ||
NM_006141.2 | NP_006132.1 | 4, 5 | 562 | |||
NR_136658.1 | 4, 5 | 768 | ||||
XM_017023008.1 | XP_016878497.1 | 4, 5 | 561 | |||
GenBank mRNA | AF035812.1 | - | 4, 5 | 537 | ||
AK291766.1 | - | 4, 5 | 541 | |||
AK295049.1 | - | 4, 5 | 561 | |||
AK304233.1 | - | 3, 4 | 417 | |||
BC025959.1 | - | 4, 5 | 537 | 4329417 | ||
BX648863.1 | - | 4, 5 | 544 | |||
DC417195.1 | - | 3, 4 | 417 | |||
EB387189.1 | - | 1, 2 | 133 |