Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000259.3 | NP_000250.3 | 2 | 330 | ||
NM_001142495.1 | NP_001135967.1 | 2 | 330 | |||
XM_005254397.3 | XP_005254454.1 | 2 | 162 | |||
XM_011521606.2 | XP_011519908.2 | 2 | 165 | |||
XM_011521607.2 | XP_011519909.2 | 2 | 165 | |||
XM_011521608.2 | XP_011519910.2 | 2 | 164 | |||
XM_011521609.2 | XP_011519911.2 | 2 | 164 | |||
XM_011521610.2 | XP_011519912.2 | 2 | 165 | |||
XM_011521611.2 | XP_011519913.2 | 2 | 164 | |||
XM_011521612.2 | XP_011519914.2 | 2 | 164 | |||
XM_017022227.1 | XP_016877716.1 | 2 | 165 | |||
GenBank mRNA | AB290180.1 | - | 2 | 89 | ||
AK307600.1 | - | 2 | 160 | |||
U90942.1 | - | 2 | 336 | |||
Y07759.1 | - | 2 | 336 |