Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TPM1 SIRNA Assays ›
Gene Symbol:
TPM1
Gene Name:
tropomyosin 1 (alpha)
Gene Aliases:
C15orf13, CMD1Y, CMH3, HEL-S-265, HTM-alpha, LVNC9, TMSA
Chromosome Location:
Chr.15: 63042196 - 63071915 on Build GRCh38
UniGene ID:
Hs.133892
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
118352
Validated
Purification
HPLC
Standard
In-Vivo Ready
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TPM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000366.5
NP_000357.3
3
446
NM_001018004.1
NP_001018004.1
3
446
NM_001018005.1
NP_001018005.1
3
446
NM_001018006.1
NP_001018006.1
3
446
NM_001018007.1
NP_001018007.1
3
446
NM_001018008.1
NP_001018008.1
2
286
NM_001018020.1
NP_001018020.1
3
446
NM_001301244.1
NP_001288173.1
3
446
NM_001301289.1
NP_001288218.1
2
286
XM_005254637.1
XP_005254694.1
4
527
XM_005254638.3
XP_005254695.2
3
889
XM_005254639.3
XP_005254696.2
3
889
XM_005254645.2
XP_005254702.2
3
889
XM_005254646.2
XP_005254703.1
2
272
XM_005254647.3
XP_005254704.1
2
272
XM_005254650.3
XP_005254707.1
2
272
XM_005254651.1
XP_005254708.1
2
272
XM_005254652.1
XP_005254709.1
2
272
XM_005254653.2
XP_005254710.1
2
272
XM_006720667.3
XP_006720730.2
3
889
XM_017022534.1
XP_016878023.1
3
889
XM_017022535.1
XP_016878024.1
3
401
XM_017022536.1
XP_016878025.1
3
889
XM_017022537.1
XP_016878026.1
3
401
XM_017022538.1
XP_016878027.1
3
889
XM_017022539.1
XP_016878028.1
3
889
XM_017022540.1
XP_016878029.1
2
272
XM_017022541.1
XP_016878030.1
2
272
GenBank mRNA
AB209041.1
-
2
323
AF474156.1
-
2
48
AK057989.1
-
4
294
AK092051.1
-
2
1163
AK092573.1
-
1
1268
AK129882.1
-
3
334
AK131384.1
-
4
460
AK298625.1
-
2
162
AK299387.1
-
3
335
AK301319.1
-
2
182
AK309170.1
-
3
655
AL050179.1
-
3
327
AY640414.1
-
3
255
AY640415.1
-
3
255
BC007433.2
-
3
396
2962925
BC050473.1
-
2
273
BC053545.1
-
2
284
5730451
BG107949.1
-
3
453
BX458841.2
-
3
315
CN423344.1
-
3
384
DQ424903.1
-
2
147
GQ472198.1
-
3
396
KU178366.1
-
3
255
KU178367.1
-
3
255
M19713.1
-
3
311
M19714.1
-
3
197
M19715.1
-
2
92
X12369.1
-
2
85
Z24727.1
-
470
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00165966_m1
Panther Classification:
Molecular Function -
actin binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
positive regulation of heart rate by epinephrine
movement of cell or subcellular component
muscle contraction
regulation of muscle contraction
cytoskeleton organization
regulation of heart contraction
regulation of cell shape
muscle filament sliding
negative regulation of cell migration
ruffle organization
positive regulation of ATPase activity
cellular response to reactive oxygen species
wound healing
sarcomere organization
positive regulation of cell adhesion
positive regulation of stress fiber assembly
ventricular cardiac muscle tissue morphogenesis
cardiac muscle contraction
negative regulation of vascular smooth muscle cell proliferation
negative regulation of vascular associated smooth muscle cell migration
actin binding
structural constituent of cytoskeleton
protein binding
cytoskeletal protein binding
structural constituent of muscle
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0