Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002471.3 | NP_002462.2 | 7 | 700 | ||
GenBank mRNA | AK309494.1 | - | 7 | 696 | ||
BC132667.1 | - | 6 | 659 | 40146689 | ||
D00943.1 | - | 5 | 629 |