Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002473.5 | NP_002464.1 | 24 | 3360 | ||
XM_011530197.2 | XP_011528499.1 | 25 | 3215 | |||
XM_017028803.1 | XP_016884292.1 | 24 | 3115 | |||
XM_017028804.1 | XP_016884293.1 | 24 | 3115 | |||
XM_017028805.1 | XP_016884294.1 | 24 | 3115 | |||
XM_017028806.1 | XP_016884295.1 | 24 | 3115 | |||
GenBank mRNA | AB191263.1 | - | 24 | 3260 | ||
AB290175.1 | - | 24 | 3153 | |||
BC049849.1 | - | 24 | 3229 | |||
CR456526.1 | - | 24 | 3161 | |||
D11393.1 | - | 3 | 351 | |||
KU159427.1 | - | 17 | 3302 | |||
M31013.1 | - | 8 | 939 | |||
M81105.1 | - | 24 | 3219 |