Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199629.1 | NP_001186558.1 | 5 | 825 | ||
NM_002475.4 | NP_002466.1 | 5 | 761 | |||
GenBank mRNA | AK304594.1 | - | 5 | 594 | ||
AK310996.1 | - | 5 | 591 | |||
BC012425.1 | - | 4 | 549 | 3862144 | ||
BC014400.1 | - | 5 | 825 | 3503030 | ||
M31211.1 | - | 4 | 529 |