Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202431.1 | NP_001189360.1 | 6 | 936 | ||
NM_002574.3 | NP_002565.1 | 6 | 1153 | |||
NM_181696.2 | NP_859047.1 | 6 | 1127 | |||
NM_181697.2 | NP_859048.1 | 6 | 933 | |||
GenBank mRNA | AK131049.1 | - | 6 | 881 | ||
BC007063.1 | - | 6 | 879 | 3961375 | ||
BC021683.1 | - | 6 | 885 | 3681912 | ||
X67951.1 | - | 6 | 872 |