Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004998.3 | NP_004989.2 | 4 | 636 | ||
GenBank mRNA | AB290177.1 | - | 4 | 619 | ||
AK291688.1 | - | 4 | 633 | |||
AL707587.1 | - | 4 | 636 | |||
BC098392.1 | - | 4 | 635 | 30527536 | ||
U14391.1 | - | 4 | 639 |