Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318245.1 | NP_001305174.1 | 43 | 7946 | ||
NM_032608.6 | NP_115997.5 | 43 | 7943 | |||
XM_011530458.2 | XP_011528760.1 | 44 | 8082 | |||
XM_011530459.2 | XP_011528761.1 | 44 | 8053 | |||
XM_011530460.2 | XP_011528762.1 | 43 | 7926 | |||
XM_011530461.2 | XP_011528763.1 | 44 | 8082 | |||
XM_011530464.2 | XP_011528766.1 | 44 | 8079 | |||
XM_011530465.2 | XP_011528767.1 | 43 | 7839 | |||
XM_017029012.1 | XP_016884501.1 | 44 | 8082 | |||
XM_017029013.1 | XP_016884502.1 | 44 | 8130 | |||
XM_017029014.1 | XP_016884503.1 | 44 | 8079 | |||
XM_017029015.1 | XP_016884504.1 | 43 | 7950 | |||
XM_017029016.1 | XP_016884505.1 | 32 | 5206 | |||
XM_017029017.1 | XP_016884506.1 | 42 | 7807 | |||
GenBank mRNA | AB042648.2 | - | 39 | 6255 | ||
AB075376.1 | - | 43 | 7943 | |||
AJ271918.1 | - | 1 | 658 | |||
AJ310931.2 | - | 43 | 7934 | |||
AK095943.1 | - | 4 | 1503 | |||
AK123936.1 | - | 4 | 1533 | |||
AL833890.1 | - | 1 | 610 | |||
AY077700.1 | - | 43 | 7856 | |||
BC144596.1 | - | 41 | 7598 | |||
BC144597.1 | - | 42 | 7740 | 9053127 | ||
BC150626.1 | - | 42 | 7740 | 9056996 | ||
CR749556.1 | - | 9 | 1989 |