Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_079420.2 | NP_524144.1 | 3 | 412 | ||
NM_079422.2 | NP_524146.1 | 3 | 222 | |||
GenBank mRNA | AK311892.1 | - | 3 | 222 | ||
AK311942.1 | - | 3 | 433 | |||
BC005318.1 | - | 3 | 222 | 3933998 | ||
CR456869.1 | - | 3 | 283 | |||
DA898482.1 | - | 3 | 438 | |||
M20643.1 | - | 3 | 199 | |||
X05451.1 | - | 3 | 204 |