Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002254.6 | NP_002245.4 | 8 | 4484 | ||
XM_005264299.2 | XP_005264356.2 | 8 | 4481 | |||
GenBank mRNA | AF035621.1 | - | 8 | 3981 | ||
BC130423.1 | - | 8 | 4055 | 40146454 | ||
BC132783.1 | - | 8 | 4055 | 40146805 | ||
BC144223.1 | - | 8 | 4049 | 9052743 | ||
BX571741.1 | - | 8 | 4405 |