Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098511.2 | NP_001091981.1 | 3 | 575 | ||
NM_001243952.1 | NP_001230881.1 | 4 | 778 | |||
NM_001243953.1 | NP_001230882.1 | 3 | 575 | |||
NM_004520.4 | NP_004511.2 | 3 | 575 | |||
GenBank mRNA | AK297347.1 | - | 3 | 321 | ||
AK302270.1 | - | 3 | 269 | |||
AK307880.1 | - | 3 | 285 | |||
AK315866.1 | - | 3 | 259 | |||
AY317140.1 | - | 1 | 88 | |||
BC031828.2 | - | 2 | 200 | 4932771 | ||
EF560716.1 | - | 3 | 286 | |||
EF560728.1 | - | 3 | 280 | |||
Y08319.1 | - | 2 | 201 |