Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002590.3 | NP_002581.2 | 3 | 3783 | ||
NM_032949.2 | NP_116567.1 | 3 | 3492 | |||
GenBank mRNA | AF061573.2 | - | 3 | 3782 | ||
AK297652.1 | - | 4 | 2361 | |||
BC036025.1 | - | 3 | 3783 | 5278690 |