Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004523.3 | NP_004514.2 | 6 | 893 | ||
GenBank mRNA | AK314258.1 | - | 6 | 796 | ||
BC126211.1 | - | 6 | 692 | 8991927 | ||
BC136474.1 | - | 6 | 692 | 9020463 | ||
U37426.1 | - | 6 | 675 | |||
X85137.1 | - | 6 | 595 |