Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012310.4 | NP_036442.3 | 9 | 1058 | ||
GenBank mRNA | AB208797.1 | - | 9 | 958 | ||
AF071592.2 | - | 9 | 975 | |||
AF179308.1 | - | 8 | 915 | |||
AF277375.1 | - | 6 | 534 | |||
AJ271784.1 | - | 8 | 921 | |||
AK022592.1 | - | 6 | 518 | |||
AK313133.1 | - | 8 | 1291 | |||
BC003664.2 | - | 9 | 996 | |||
BC038459.1 | - | 9 | 995 | |||
BC049218.1 | - | 9 | 964 | |||
BC050548.1 | - | 9 | 957 | 4538604 |