Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF20B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284259.1 | NP_001271188.1 | 2 | 202 | ||
NM_016195.3 | NP_057279.2 | 2 | 202 | |||
GenBank mRNA | AB033337.1 | - | 2 | 155 | ||
AK074336.1 | - | 2 | 153 | |||
AK290439.1 | - | 2 | 182 | |||
AL117496.1 | - | 2 | 157 | |||
AY282406.1 | - | 2 | 179 | |||
BC042130.1 | - | 2 | 154 | |||
BC046134.1 | - | 2 | 155 | |||
BC058913.1 | - | 2 | 158 | |||
BC093089.1 | - | 2 | 157 | |||
BC108688.1 | - | 2 | 164 | |||
CB130618.1 | - | 2 | 202 |