Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF20B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284259.1 | NP_001271188.1 | 5, 6 | 590 | ||
NM_016195.3 | NP_057279.2 | 5, 6 | 590 | |||
GenBank mRNA | AB033337.1 | - | 5, 6 | 543 | ||
AK074336.1 | - | 5, 6 | 541 | |||
AK290439.1 | - | 5, 6 | 570 | |||
AL117496.1 | - | 5, 6 | 545 | |||
AY282406.1 | - | 5, 6 | 567 | |||
BC042130.1 | - | 5, 6 | 542 | |||
BC046134.1 | - | 5, 6 | 543 | |||
BC058913.1 | - | 5, 6 | 546 | |||
BC093089.1 | - | 5, 6 | 545 | |||
BC108688.1 | - | 5, 6 | 552 |