Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134775.1 | NP_001128247.1 | 3 | 653 | ||
NM_001134776.1 | NP_001128248.1 | 3 | 532 | |||
NM_001318734.1 | NP_001305663.1 | 3 | 590 | |||
NM_022822.2 | NP_073733.1 | 3 | 627 | |||
XM_005274183.1 | XP_005274240.1 | 3 | 569 | |||
XM_005274184.1 | XP_005274241.1 | 3 | 590 | |||
GenBank mRNA | AK022449.1 | - | 3 | 519 | ||
AK022907.1 | - | 3 | 572 | |||
AK126489.1 | - | 1 | 1011 | |||
AK315725.1 | - | 3 | 522 | |||
AL136864.1 | - | 3 | 553 | |||
BC034373.1 | - | 3 | 515 | 4342879 | ||
DA198727.1 | - | 3 | 475 |