Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134774.1 | NP_001128246.1 | 15 | 2014 | ||
NM_001134775.1 | NP_001128247.1 | 16 | 2308 | |||
NM_001134776.1 | NP_001128248.1 | 16 | 2187 | |||
NM_001318734.1 | NP_001305663.1 | 16 | 2245 | |||
NM_022822.2 | NP_073733.1 | 16 | 2282 | |||
XM_005274183.1 | XP_005274240.1 | 16 | 2224 | |||
XM_017018133.1 | XP_016873622.1 | 14 | 2181 | |||
GenBank mRNA | AK022449.1 | - | 16 | 2174 | ||
AK022907.1 | - | 16 | 2227 | |||
AK094593.1 | - | 14 | 1884 | |||
AK126489.1 | - | 13 | 3047 | |||
AL136864.1 | - | 16 | 2208 | |||
BC034373.1 | - | 16 | 2170 | 4342879 |