Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031217.3 | NP_112494.3 | 2 | 475 | ||
XM_017018379.1 | XP_016873868.1 | 2 | 1031 | |||
GenBank mRNA | AB062483.1 | - | 2 | 376 | ||
AK312576.1 | - | 2 | 420 | |||
AL136819.1 | - | 2 | 422 | |||
AY791349.1 | - | 2 | 405 | |||
BC026090.1 | - | 2 | 455 | |||
BC048347.2 | - | 1 | 340 | 5576803 |