Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130107.1 | NP_001123579.1 | 2 | 439 | ||
NM_005552.4 | NP_005543.2 | 2 | 439 | |||
NM_182923.3 | NP_891553.2 | 2 | 439 | |||
GenBank mRNA | AB451299.1 | - | 1 | 119 | ||
AB451431.1 | - | 1 | 119 | |||
AF527736.1 | - | 2 | 389 | |||
AK294508.1 | - | 2 | 424 | |||
AY180164.2 | - | 2 | 386 | |||
BC008881.2 | - | 2 | 400 | 4301579 | ||
BP274336.1 | - | 2 | 439 | |||
DA501532.1 | - | 2 | 427 | |||
L04733.1 | - | 2 | 395 |