Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF26A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015656.1 | NP_056471.1 | 4, 5 | 931 | ||
GenBank mRNA | AB033062.2 | - | 3, 4 | 805 | ||
AB290161.1 | - | 3, 4 | 901 |