Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000029.3 | NP_000020.1 | 5 | 2034 | ||
GenBank mRNA | AB209769.1 | - | 5 | 1546 | ||
AK222798.1 | - | 5 | 1565 | |||
AK222815.1 | - | 5 | 1566 | |||
AK293507.1 | - | 6 | 1319 | |||
AK303755.1 | - | 6 | 1507 | |||
BC011519.1 | - | 5 | 1554 | 4213559 | ||
K02215.1 | - | 5 | 1565 |