Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKN1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000389.4 | NP_000380.1 | 3 | 1558 | ||
NM_001220777.1 | NP_001207706.1 | 3 | 1503 | |||
NM_001220778.1 | NP_001207707.1 | 3 | 1668 | |||
NM_001291549.1 | NP_001278478.1 | 4 | 1710 | |||
NM_078467.2 | NP_510867.1 | 4 | 1505 | |||
GenBank mRNA | AB209881.1 | - | 3 | 1515 | ||
BC000275.1 | - | 3 | 1498 | 3355833 | ||
BC000312.2 | - | 3 | 1652 | 2821049 | ||
BC001935.1 | - | 3 | 1513 | 2823668 | ||
BC013967.2 | - | 3 | 1516 | 2822909 | ||
BQ941282.1 | - | 1 | 103 | |||
L25610.1 | - | 3 | 1511 | |||
L26165.1 | - | 3 | 1511 | |||
U03106.1 | - | 3 | 1509 | |||
U09579.1 | - | 3 | 1527 |